missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-392B
This item is not returnable.
View return policy
Description
DNMT1 Polyclonal antibody specifically detects DNMT1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DNMT1 | |
| Polyclonal | |
| Western Blot | |
| AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
| The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human DNA-Methyltranserase 1 using the numbering given in entry NP_001370.1 (GeneID 1786). | |
| 0.1 mL | |
| Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators | |
| 1786 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction