missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT1(phospho S714) & DNMT1 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DP0018.1Set
This item is not returnable.
View return policy
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the DNMT1 protein, and the other against the specific S714 phosphorylated site of DNMT1 for use in in situ Proximity Ligation Assay.Specifications
| DNMT1 | |
| Unconjugated | |
| AIM/CXXC9/DNMT/FLJ16293/MCMT/MGC104992 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-DNMT1 S714 rabbit polyclonal antibody (20 ul) In PBS (0.09% (w/v) sodium azide) 2. DNMT1 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| DNA (cytosine-5-)-methyltransferase 1 | |
| DNMT1 | |
| RUO | |
| 1786 | |
| Human | |
| Ab Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction