missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dnmt2 Antibody (102B1259.2) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB200-587JF635
This item is not returnable.
View return policy
Description
Dnmt2 Monoclonal antibody specifically detects Dnmt2 in Human,Mouse,Zebrafish samples. It is validated for Flow Cytometry,Western Blot
Specifications
| Dnmt2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1 | |
| This antibody was raised against synthetic peptides corresponding to mouse Dnmt2. | |
| 0.1 mL | |
| Chromatin Research, Epigenetics | |
| 1787 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Western Blot | |
| 102B1259.2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction