missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3A Antibody (64B814.1), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56521AF405
This item is not returnable.
View return policy
Description
DNMT3A Monoclonal specifically detects DNMT3A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| DNMT3A | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 64B814.1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Q9Y6K1 | |
| DNMT3A | |
| This DNMT3A Antibody (64B814.1) was raised against bacteria expressed HIS-tag recombinant mouse Dnmt3a. | |
| 0.1 mL | |
| Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
| 1788 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction