missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNP Monoclonal Antibody (LO-DNP-30)

Rat Monoclonal Antibody
Brand: Invitrogen™ 048888
This item is not returnable.
View return policy
Description
DNP Monoclonal Antibody for IHC, ELISA
DNP (2,4-dinitrophenol) is a toxic dye, chemically related to trinitrophenol (picric acid). It is used in biochemical studies of oxidative processes where it uncouples oxidative phosphorylation and is also used as a metabolic stimulant. 2,4-Dinitrophenol is used in the manufacture of dyes, wood preservatives, and as a pesticide. The acute (short-term) effects of 2,4-dinitrophenol in humans through oral exposure are nausea, vomiting, sweating, dizziness, headaches, and loss of weight. Chronic (long-term) oral exposure to 2,4-dinitrophenol in humans has resulted in the formation of cataracts and skin lesions, weight loss, and has caused effects on the bone marrow, central nervous system (CNS), and cardiovascular system. [National Center for Biotechnology Information. PubChem Compound Database; CID=1493].
Specifications
| DNP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Rat | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgE |
| ELISA, Immunohistochemistry | |
| LO-DNP-30 | |
| Unconjugated | |
| C6H3N2O4; C6H4N2O5; Dinitrophenol; DNP | |
| Synthetic DNP. | |
| 200 μg | |
| Primary | |
| -20°C | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction