Learn More
DNTTIP1 Monoclonal Antibody (OTI1F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501132
This item is not returnable.
View return policy
Description
DNTTIP1 binds DNA and enhances the activity of terminal deoxynucleotidyltransferase, a DNA polymerase that catalyzes the polymerization of DNA in the absence of a DNA template.Specifications
| DNTTIP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H147 | |
| DNTTIP1 | |
| Full length human recombit protein of human DNTTIP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F4 | |
| Unconjugated | |
| DNTTIP1 | |
| C20orf167, Tdif1, dJ447F3.4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 116092 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |