Learn More
DNTTIP1 Monoclonal Antibody (OTI4D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501131S
Encompass_Preferred
 View more versions of this product
Product Code. 17533381
  • € 242.00 / 30µL
Description
DNTTIP1 Monoclonal antibody specifically detects DNTTIP1 in Human samples. It is validated for Western Blot
Specifications
Specifications
DNTTIP1
Monoclonal
0.79 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H147
Mouse
Affinity chromatography
RUO
116092
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI4D1
Unconjugated
DNTTIP1
6430706C13Rik; C20orf167; deoxynucleotidyltransferase; deoxynucleotidyltransferase terminal interacting protein 1; deoxynucleotidyltransferase terminal-interacting protein 1; deoxynucleotidyltransferase, terminal, interacting protein 1; dJ447F3.4; Dnttip1; RP3-447F3.5; Tdif1; TdT binding protein; tdT-interacting factor 1; terminal deoxynucleotidyltransferase interacting factor 1; terminal deoxynucleotidyltransferase-interacting factor 1
Full length human recombinant protein of human DNTTIP1 produced in HEK293T cell.