missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DOC2B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15738101 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15738101 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15738101 Supplier Invitrogen™ Supplier No. PA560265

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LKKLKPNHTK TFSICLEKQL PVDKTEDKSL EE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

There are at least two protein isoforms of the Double C2 protein, namely alpha and beta, which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2B is expressed ubiquitously and is suggested to be involved in Ca-dependent intracellular vesicle trafficking in various types of cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOC2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOC2B
Gene Accession No. Q14184
Gene Alias doc2b; doc2-beta; DOC2BL; double C2 domain beta; double C2 protein beta; double C2, beta; double C2-like domain-containing protein beta; double C2-like domain-containing protein beta; LOW QUALITY PROTEIN: double C2-like domain-containing protein beta; double C2-like domains, beta; OTTMUSP00000006522
Gene Symbols DOC2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DOC2B. Recombinant protein control fragment (Product #RP-100079).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8447
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.