missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK10 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60669R
This item is not returnable.
View return policy
Description
DOCK10 Polyclonal antibody specifically detects DOCK10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DOCK10 | |
| Polyclonal | |
| Western Blot | |
| dedicator of cytokinesis 10, dedicator of cytokinesis protein 10, DKFZp781A1532, DRIP2, KIAA0694protein zizimin 3, Nbla10300, ZIZ3dopamine receptor interacting protein 2, zizimin3, zizimin-3 | |
| The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human dedicator of cytokinesis 10 using the numbering given in entry NP_055504.2 (GeneID 55619). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55619 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction