missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK7 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78749PCP
This item is not returnable.
View return policy
Description
DOCK7 Polyclonal antibody specifically detects DOCK7 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| DOCK7 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| dedicator of cytokinesis 7, dedicator of cytokinesis protein 7, KIAA1771ZIR2 | |
| The immunogen for this product maps to a region between residue 2090 and 2140 of human Dedicator of Cytokinesis 7 using the numbering given in entry Q96N67.4 (GeneID 85440). | |
| 0.1 mL | |
| Cell Biology, GPCR, Neuroscience, Signal Transduction | |
| 85440 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction