missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOCK7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109653
This item is not returnable.
View return policy
Description
Immunogen sequence: VSAVPEESEM DPHVRDCIRS YTEDWAIVIR KYHKLGTGFN PNTLDKQKER QKGLPKQVFE SDEAPDGNSY QDDQDDLKRR SMSIDDTPRG SWACSIFDLK N.
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) that plays a role in axon formation and neuronal polarization. The encoded protein displays GEF activity toward RAC1 and RAC3 Rho small GTPases but not toward CDC42. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| DOCK7 | |
| Polyclonal | |
| Unconjugated | |
| DOCK7 | |
| 3110056M06Rik; dedicator of cytokinesis 7; dedicator of cytokinesis protein 7; Dock7; EIEE23; Gm430; KIAA1771; m; protein moonlight; ZIR2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 313388, 67299, 85440 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.49 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8R1A4, Q96N67 | |
| DOCK7 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human DOCK7 (NP_0013544901). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction