Learn More
Invitrogen™ DOCK7 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103779
Product Code. 16600235
  • € 494.00 / 100µL
Description
Antibody detects endogenous levels of total DOCK7.
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) that plays a role in axon formation and neuronal polarization. The encoded protein displays GEF activity toward RAC1 and RAC3 Rho small GTPases but not toward CDC42. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DOCK7
Polyclonal
Unconjugated
DOCK7
3110056M06Rik; dedicator of cytokinesis 7; dedicator of cytokinesis protein 7; Dock7; EIEE23; Gm430; KIAA1771; m; protein moonlight; ZIR2
Rabbit
Affinity chromatography
RUO
313388, 67299, 85440
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8R1A4, Q96N67
DOCK7
A synthesized peptide derived from human DOCK7(Accession Q96N67), corresponding to amino acid residues D197-P247.
100 μL
Primary
Human, Mouse, Rat