Learn More
DOCK7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13000-1-AP-150UL
 View more versions of this product
Product Code. 16811004
  • € 410.00 / 150µL
Description
Multiple isoforms of DOCK 7 exist due to alternative splicing events. The DOCK 7 antibody (13000-1-AP) can recognize all isoforms.
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) that plays a role in axon formation and neuronal polarization. The encoded protein displays GEF activity toward RAC1 and RAC3 Rho small GTPases but not toward CDC42. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
DOCK7
Polyclonal
Unconjugated
DOCK7
dedicator of cytokinesis 7, DOCK7, KIAA1771, ZIR2
Rabbit
Antigen Affinity Chromatography
RUO
313388, 67299, 85440
-20°C
Liquid
Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R1A4, Q96N67
DOCK7
DOCK7 Fusion Protein Ag3671
150 μL
Primary
Human, Mouse, Rat