missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK8 Monoclonal Antibody (OTI5C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506495
This item is not returnable.
View return policy
Description
The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.Specifications
| DOCK8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NF50 | |
| DOCK8 | |
| Human recombit protein fragment corresponding to amino acids 833-1160 of human DOCK8 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5C4 | |
| Unconjugated | |
| DOCK8 | |
| HEL-205, MRD2, ZIR8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 81704 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?