missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK8 Monoclonal Antibody (OTI5C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506495S
This item is not returnable.
View return policy
Description
DOCK8 Monoclonal antibody specifically detects DOCK8 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| DOCK8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NF50 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 81704 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI5C4 | |
| Unconjugated | |
| Dock8 | |
| 1200017A24Rik; 5830472H07Rik; A130095G14Rik; AI461977; dedicator of cytokinesis 8; dedicator of cytokinesis protein 8; Dock8; epididymis luminal protein 205; HEL-205; MRD2; ZIR8 | |
| Human recombinant protein fragment corresponding to amino acids 833-1160 of human DOCK8 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction