missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK9 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-264C
This item is not returnable.
View return policy
Description
DOCK9 Polyclonal antibody specifically detects DOCK9 in Human samples. It is validated for Western Blot
Specifications
| 50mM Sodium Borate | |
| RUO | |
| Human |
| Immunogen affinity purified | |
| 23348 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction