missing translation for 'onlineSavingsMsg'
Learn More

Dodecanonitrile, 98%, Thermo Scientific™

Product Code. 11489336
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489336 250 g 250g
11469336 10 g 10g
11479336 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489336 Supplier Thermo Scientific Alfa Aesar Supplier No. A19038.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Dodecanonitrile in xylene is used as a solvent and in the presence of sulfur was reduced with molybdenum sulfide catalyst yielded 70-5% dodecylmercaptan.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2437-25-4
Molecular Formula C12H23N
Molecular Weight (g/mol) 181.32
MDL Number MFCD00001946
InChI Key VXCUURYYWGCLIH-UHFFFAOYSA-N
Synonym lauronitrile, undecyl cyanide, dodecanonitrile, 1-cyanoundecane, n-undecyl cyanide, lauric nitrile, decylacetonitrile, lauric acid nitrile, dodecane nitrile, nitrile 12
PubChem CID 17092
IUPAC Name dodecanenitrile
SMILES CCCCCCCCCCCC#N

Specifications

Melting Point 4°C
Density 0.83
Boiling Point 272°C to 274°C
Flash Point 114°C (237°F)
Refractive Index 1.436
Quantity 250 g
Beilstein 970348
Solubility Information Insoluble in water.
Formula Weight 181.32
Percent Purity 98%
Chemical Name or Material Dodecanonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.