missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DOG-1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16546301
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16546301 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16546301 Supplier Invitrogen™ Supplier No. PA587947

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: LO2, HeLa, BT-474, Mouse lung, Mouse eye, Mouse kidney, Rat liver, Rat skeletal muscle; Cellular Location: Cell membrane, Cytoplasm, Multi-pass membrane protein Immunogen sequence: ASHLFDNPAT VFFSVFMALW AATFMEHWKR KQMRLNYRWD LTGFEEEEEA VKDHPRAEYE ARVLEKSLKK ESRNKEKRRH IPEESTNKWK QRVKTAMAGV K.

DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOG-1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANO1
Gene Accession No. Q5XXA6
Gene Alias ANO1; anoctamin 1; anoctamin 1, calcium activated chloride channel; anoctamin-1; Ca2+-activated Cl- channel; calcium activated chloride channel; discovered on gastrointestinal stromal tumors protein 1; DOG1; FLJ10261; oral cancer overexpressed 2; Oral cancer overexpressed protein 2; ORAOV2; TAOS2; Tmem16a; Transmembrane protein 16A; transmembrane protein 16A (eight membrane-spanning domains); tumor-amplified and overexpressed sequence 2
Gene Symbols ANO1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-500 of human ANO1 (NP_0605135).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55107
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.