missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92548-0.02ml
This item is not returnable.
View return policy
Description
DOK2 Polyclonal antibody specifically detects DOK2 in Human samples. It is validated for Western Blot
Specifications
| DOK2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| docking protein 2, docking protein 2, 56kD, docking protein 2, 56kDa, Dok-2, Downstream of tyrosine kinase 2, p56(dok-2), P56DOK, p56dok-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DOK2 (NP_003965.2). MGDGAVKQGFLYLQQQQTFGKKWRRFGASLYGGSDCALARLELQEGPEKPRRCEAARKVIRLSDCLRVAEAGGEASSPRDTSAFFLETKERLYLLAAPAAERGDWVQAICLLAFPGQRKELSGPEGKQSRPCMEENELYSSAVTVGPHKEFAVTMRPTEASERCHLRGSYTLRAGESALELWGGPEPGTQLYDWPYRFLRRFGRDKVTFSFEAGRRCVSGEGNFEFETRQGNEIFLALEEAISAQKNAAPATPQPQPATI | |
| 0.02 mL | |
| Signal Transduction | |
| 9046 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction