missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK7 Monoclonal Antibody (OTI2C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504787
This item is not returnable.
View return policy
Description
The protein encoded by this gene is essential for neuromuscular synaptogenesis. The protein functions in aneural activation of muscle-specific receptor kinase, which is required for postsynaptic differentiation, and in the subsequent clustering of the acetylcholine receptor in myotubes. This protein can also induce autophosphorylation of muscle-specific receptor kinase. Mutations in this gene are a cause of familial limb-girdle myasthenia autosomal recessive, which is also known as congenital myasthenic syndrome type 1B. Alternative splicing results in multiple transcript variants.Specifications
| DOK7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q18PE1 | |
| DOK7 | |
| Full length human recombit protein of human DOK7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI2C12 | |
| Unconjugated | |
| DOK7 | |
| C4orf25, CMS10, CMS1B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 285489 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction