Learn More
DOK7 Monoclonal Antibody (OTI4C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504786S
Encompass_Preferred
 View more versions of this product
Product Code. 17522422
  • € 242.00 / 30µL
Description
DOK7 Monoclonal antibody specifically detects DOK7 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
DOK7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NUV9
Mouse
Affinity chromatography
RUO
55303
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Western Blot
OTI4C6
Unconjugated
GIMAP4
A930013K19Rik; AW049091; C4orf25; CMS10; CMS1B; docking protein 7; DOK7; Dok-7; Downstream of tyrosine kinase 7; EF-12; Oit5; oncoprotein induced transcript 5; protein Dok-7; RGD1566416
Full length human recombinant protein of human GIMAP4 produced in HEK293T cell.
30 μL