missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, PE/Cy5.5 (Pre-adsorbed), Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | PE-Cyanine5.5 |
| Formulation | PBS |
| Host Species | Donkey |
| Immunogen | Goat IgG whole molecule |
| Purification Method | Multi-step |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?