missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Rabbit IgG Fc Secondary Antibody, FITC, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-73255
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Rabbit samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifications
| IgG Fc | |
| Polyclonal | |
| FITC | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Rabbit IgG F(c) fragment | |
| 2 mg | |
| Adaptive Immunity, Immunology | |
| Reconstitute with 1.0 mL deionized water (or equivalent). | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
| LYOPH | |
| Western Blot 1:2000 - 1:20000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:100000 | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction