missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Sheep IgG (H+L) Secondary Antibody, Texas Red, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NB7582
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Texas Red | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Sheep IgG whole molecule | |
| 2 mg | |
| Adaptive Immunity, Immunology | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Donkey Serum, Sheep IgG and Sheep Serum. | |
| Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction