missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Door-Open-Recognition for UN750plus

Product Code. 17560456 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN750plus Oven
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17560456

Brand: Memmert™ UN750plusV5

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Door-Open-Recognition for Memmert Ovens.

  • Only for units with TwinDISPLAY

Specifications

For Use With (Equipment) UN750plus Oven
Type Door Open Recognition
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.