Learn More
Memmert™ Door with Safety Lock for UN30
Product Code. 17590066 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
Product Code. 17590066
Brand: Memmert™ UN30B6
Only null left
Add to basket
Add to basket
Door with lock and key for use with Memmert Ovens.
  • With lock and key
  • Accessory
Specifications
For Use With (Equipment) UN30 Oven
Type Door with Safety Lock