missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-52742
This item is not returnable.
View return policy
Description
Dopamine Polyclonal antibody specifically detects Dopamine in Human, Rat samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Dopamine | |
| Polyclonal | |
| ELISA 1:1000 - 1:10000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:2000 | |
| 3 4 dihydroxyphenethylamine, 3 hydroxytyramine, DA, Dopamine, Oxytyramine | |
| BSA-Dopamine | |
| 100 μg | |
| Neuroscience | |
| Lyophilized powder, reconstituted in distilled water. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| Tris 0, 1 M, glycine 0, 1M, sucrose 2% | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction