missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Dopamine beta-Hydroxylase Antibody (DBH/7222), Novus Biologicals™
Mouse Monoclonal Antibody
€ 245.00 - € 486.00
Specifications
| Antigen | Dopamine beta-Hydroxylase |
|---|---|
| Clone | DBH/7222 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043655
|
Novus Biologicals™
NBP3-26770-20ug |
20 μg |
€ 245.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043845
|
Novus Biologicals™
NBP3-26770-100ug |
100 μg |
€ 486.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dopamine beta-Hydroxylase Monoclonal antibody specifically detects Dopamine beta-Hydroxylase in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Dopamine beta-Hydroxylase | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Lipid and Metabolism, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 1621 | |
| IgG2 κ | |
| Protein A or G purified |
| DBH/7222 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DBM, dopamine beta-hydroxylase, dopamine beta-hydroxylase (dopamine beta-monooxygenase), Dopamine beta-monooxygenase, EC 1.14.17.1 | |
| Recombinant fragment (around aa200-400) of human Dopamine beta-Hydroxylase protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title