missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine beta-Hydroxylase Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB300-112
This item is not returnable.
View return policy
Description
Dopamine beta-Hydroxylase Polyclonal specifically detects Dopamine beta-Hydroxylase in Human, Primate samples. It is validated for Western Blot.
Specifications
| Dopamine beta-Hydroxylase | |
| Polyclonal | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| DBM, dopamine beta-hydroxylase, dopamine beta-hydroxylase (dopamine beta-monooxygenase), Dopamine beta-monooxygenase, EC 1.14.17.1 | |
| Sheep | |
| 75 kDa | |
| 0.1 mL | |
| Primary | |
| Specific for the ∼75k DBH protein in Western blots. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.4 mg/mL | |
| Western Blot 1:1000 | |
| P09172 | |
| DBH | |
| Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH. Accession # P09172 | |
| Antigen Affinity-purified | |
| RUO | |
| 1621 | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction