missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D1R/DRD1 Antibody (887438), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8276H
This item is not returnable.
View return policy
Description
Dopamine D1R/DRD1 Monoclonal antibody specifically detects Dopamine D1R/DRD1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Dopamine D1R/DRD1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 887438 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A | |
| NS0 mouse myeloma cell line transfected with human Dopamine D1 R/DRD1, Met1-Thr446, Accession # P21728 | |
| 0.1 mL | |
| GPCR, Neuroscience, Neurotransmission | |
| 1812 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction