missing translation for 'onlineSavingsMsg'
Learn More

Dopamine-d3 Hydrochloride, TRC

Product Code. 30377133
Change view
Click to view available options
Quantity:
1.5 mg
3 mg
15 mg
Unit Size:
1.5mg
3mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377133 1.5 mg 1.5mg
30374576 3 mg 3mg
30371785 15 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30377133 Supplier Toronto Research Chemicals Supplier No. TRCD5337851.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 53587-30-7
Molecular Formula C8 D3 H8 N O2 . Cl H
Molecular Weight (g/mol) 192.66
Synonym 2-(3,4-Dihydroxyphenyl-d3)ethylamine HCl
IUPAC Name 4-(2-aminoethyl)-3,5,6-trideuteriobenzene-1,2-diol;hydrochloride
SMILES Cl.[2H]c1c([2H])c(CCN)c([2H])c(O)c1O

Spezifikation

Quantity 1.5 mg
InChI Formula InChI=1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H/i1D,2D,5D;
Formula Weight 192.0745
Percent Purity >95%
Chemical Name or Material Dopamine-d3 Hydrochloride
Grade HPLC
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.