missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doppel Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97004AF350
This item is not returnable.
View return policy
Description
Doppel Polyclonal antibody specifically detects Doppel in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Doppel | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| dJ1068H6.4, DOPPEL, DPLprion gene complex, downstream, MGC41841, Prion protein 2, prion protein 2 (dublet), PrPLPprion-like protein doppel | |
| Produced in rabbits immunized with purified, recombinant Human Doppel (Accession#: NP_036541.2; Met1-Gly152) | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 23627 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction