missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doramectin, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Doramectin |
| CAS | 117704-25-3 |
| CAS Max % | ≤ 100% |
| Linear Formula | C50H74O14 |
| Color | White |
| Melting Point | 160.5°C to 162.2°C |
| Synonym | 25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)avermectin |
| Molecular Weight (g/mol) | 899.11 g/mol |
| Quantity | 100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction