missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dorsomorphin, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Dorsomorphin (Compound C) is a selective and ATP-competitive AMPK inhibitor (Ki=109 nM in the absence of AMP). Dorsomorphin (BML-275) selectively inhibits BMP type I receptors ALK2, ALK3, and ALK6. Dorsomorphin induces autophagy.
Specifications
Specifications
| Chemical Name or Material | Dorsomorphin |
| CAS | 866405-64-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H25N5O |
| Quantity | 10 mg |
| Synonym | Compound C BML-275 |
| Solubility Information | 1M HCl : 50 mg/mL (125.16 mM; ultrasonic and adjust pH to 1 with HCl) ∣DMSO : 5 mg/mL (12.52 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : 3.33 mg/mL (8.34 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | C12=C(C3=CC=NC=C3)C=NN1C=C(C4=CC=C(OCCN5CCCCC5)C=C4)C=N2 |
| Molecular Weight (g/mol) | 399.49 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction