missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOT1L Antibody (OTI1D8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46473
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
DOT1L Monoclonal specifically detects DOT1L in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| DOT1L | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1D8 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| DOT1L | |
| Human recombinant protein fragment corresponding to amino acids 1-434 of human DOT1L (NP_115871) produced in E.coli. | |
| 0.1 mL | |
| Chromatin Research, Stem Cell Markers | |
| 84444 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction