missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOT1L Antibody (OTI1D8), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72297PCP
This item is not returnable.
View return policy
Description
DOT1L Monoclonal antibody specifically detects DOT1L in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| DOT1L | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1D8 | |
| Western Blot, Immunohistochemistry | |
| DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4 | |
| Human recombinant protein fragment corresponding to amino acids 1-434 of human DOT1L (NP_115871) produced in E.coli. | |
| 0.1 mL | |
| Chromatin Research, Stem Cell Markers | |
| 84444 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction