Learn More
DOT1L Mouse anti-Human, Clone: OTI3E9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF802740
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DOT1L encodes histone methyltransferase, and methylates ′Lys-79′ of histone H3. Nucleosomes are preferred as substrate compared to free histones, binds to DNA.Specifications
| DOT1L | |
| Monoclonal | |
| Unconjugated | |
| DOT1L | |
| DOT1, KMT4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84444 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3E9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8TEK3 | |
| DOT1L | |
| Human recombit protein fragment corresponding to amino acids 1008-1424 of human DOT1L produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.