Learn More
Wesbart™ Double Platform Test Tube Rack
Product Code. 15815781
Change view
Click to view available options
Positions:
25
40
50
54
100
145
Holds:
Tubes (14 mm)
Tubes (15 mL), Tubes (17 mm)
Tubes (18 mm)
Tubes (20 mm)
Tubes (30 mm)
Tubes (5 mL)
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Holds unitSize
15815781 145 Tubes (15 mL), Tubes (17 mm) Each
11781704 50 Tubes (14 mm) Each
15713368 25 Tubes (18 mm) Each
11711714 100 Tubes (18 mm) Each
15723368 25 Tubes (30 mm) Each
11731714 40 Tubes (20 mm) Each
15751589 54 Tubes (5 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15815781 Supplier Wesbart™ Supplier No. DP17145HG
Only null left
Add to basket
Add to basket
Test Tube racks are an effective way of storing Falcon/ Universal tubes, Bijous, cryogenic vials or any size glass bottles.
  • Racks are available in aluminum and stainless steel with the option of a hand grip
  • Test Tube racks can be designed to fit in horizontal cold store or be free standing
  • Different size test tubes racks are available to suit your requirements
  • Labels can be fitted to racks so they can be easily identified
Specifications
Holds Tubes (15 mL), Tubes (17 mm)
Material Aluminum
Positions 145
For Use With (Equipment) Horizontal outer AH949/2NM
Depth (Metric) 672 mm
Diameter (Metric) Opening 17 mm
Product Type Double Platform Test Tube Rack
Height (Metric) 70 mm
Width (Metric) 133 mm