Learn More
SI Analytics™ Double-Platinum Combination Electrode
Ideal for measurements that take place in small receptables. SI Analytics™ Double-Platinum Combination Electrode is are specially designed electrodes for the life science and pharmacy industries.
Brand:  SI Analytics™ 285102060
Additional Details : Weight : 0.10000kg
Product Code. 12665706
  • € 260.00 / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
  • Pharmacy and all Life Science sectors offer the most application areas for Microelectrodes
  • Ideal for measurement that often takes place in small receptacles as i.e. ampoules, vials, small vessels, NMR tubes etc.
  • Due to this small sample volume it is important to take care of the electrode's diameter and the minimum immersion length in order to place the junction into the sample as well
  • Developed especially for these circumstances and hence are spcialized in these applications
  • They are the specialists for Life Science and Pharmacy
Specifications
Specifications
Glass
Coulometric Titration Vessels TZ 1751 and TZ 1754
KF 1150
Double Platinum Electrode
Fixed Cable
Taper NS 7.5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers