Learn More
Schneider Gerd™ Double Spatula
Use this spatula for a variety of pharmaceutical applications. Schneider Gerd™ Double Spatulas are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 8305
Additional Details : Weight : 0.00900kg
 View more versions of this product
Product Code. 10051031
  • € 4.10 / Each
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
Description
Description
  • Spatulas are ideal for working with powders, stirring and mixing
  • Two end shapes offer increased versatility
  • Perfect for miniscule amounts of chemicals
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
1 Pk.
11 mm
250 mm
Double Ended Spatula
250 x 11 mm