missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doublecortin Antibody (3E1), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92684AF700
This item is not returnable.
View return policy
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Doublecortin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| O43602 | |
| DCX | |
| Recombinant full length human Lis-A isoform of Doublecortin purified from E. coli. [UniProt# O43602] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Cow, Pig and other Mammals. | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 30 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin) | |
| Mouse | |
| 49.3 kDa | |
| 0.1 mL | |
| Neuroscience, Phospho Specific, Signal Transduction | |
| 1641 | |
| Human, Mouse, Rat, Pig, Bovine, Equine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction