Learn More
Gilac™ Dough Container
Product Code. p-200009582
Click to view available options
Volume (Metric):
9 L
14 L
15 L
23 L
Color:
Blue
Gray
Green
Red
Translucent
White
Yellow
Dimensions (L x W x H):
600 x 400 x 55 mm
600 x 400 x 76 mm
600 x 400 x 90 mm
600 x 400 x 130 mm
Unit Size:
Each
Product Code. 17988491
Brand: Gilac™ G179431
Only null left
Add to basket
Add to basket
Dough Container is ideal for dough, pastries, chocolate molds storage, with rounded corners and bottom for easy cleaning.
  • 600 x 400 dough container, ideal for dough, pastries, chocolate molds... storage.
  • Stacking with and without lid, maximum stacking height capacity.
  • Innovative design for even greater convenience:
    • Rounded corners and bottom for easy cleaning.
    • Vertical sides without inner edge avoids marking the dough.
    • The entire surface of the container is usable.
    • Completely flat bottom means dough detaches easily, no loss, no food deposit left.
    • Optimised side reinforcements allows you to customise containers with your logo.
    • Rounded outside corners guarantee a damage-free working environment.
  • Translucent container facilitates the control of the contents.
    • Allows you to monitor dough rising without lifting the container.
Specifications
Product Type Dough Container
Material Polyethylene
Color White
Dimensions (L x W x H) 600 x 400 x 55 mm
Volume (English) 2.4 gal.
Volume (Metric) 9 L