Learn More
Gilac™ Dough Container
Click to view available options
Volume (Metric):
9 L
14 L
15 L
23 L
Color:
Blue
Gray
Green
Red
Translucent
White
Yellow
Dimensions (L x W x H):
600 x 400 x 55 mm
600 x 400 x 76 mm
600 x 400 x 90 mm
600 x 400 x 130 mm
Unit Size:
Each
Description
- 600 x 400 dough container, ideal for dough, pastries, chocolate molds... storage.
- Stacking with and without lid, maximum stacking height capacity.
- Innovative design for even greater convenience:
- Rounded corners and bottom for easy cleaning.
- Vertical sides without inner edge avoids marking the dough.
- The entire surface of the container is usable.
- Completely flat bottom means dough detaches easily, no loss, no food deposit left.
- Optimised side reinforcements allows you to customise containers with your logo.
- Rounded outside corners guarantee a damage-free working environment.
- Translucent container facilitates the control of the contents.
- Allows you to monitor dough rising without lifting the container.
Specifications
Specifications
| Product Type | Dough Container |
| Material | Polyethylene |
| Color | White |
| Dimensions (L x W x H) | 600 x 400 x 130 mm |
| Volume (English) | 6.1 gal. |
| Volume (Metric) | 23 L |