missing translation for 'onlineSavingsMsg'
Learn More

Dovitinib lactate, MedChemExpress

Product Code. 16421546
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16421546

Brand: Medchem Express HY10207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dovitinib lactate (TKI258 lactate) is a multi-targeted tyrosine kinase inhibitor with IC50s of 1, 2, 8/9, 10/13/8, 27/210 nM for FLT3, c-Kit, FGFR1/3, VEGFR1/2/3 and PDGFRα/β, respectively.

Dovitinib lactate (TKI258 lactate) is a multi-targeted tyrosine kinase inhibitor with IC50s of 1, 2, 8/9, 10/13/8, 27/210 nM for FLT3, c-Kit, FGFR1/3, VEGFR1/2/3 and PDGFRα/β, respectively.

Specifications

Chemical Name or Material Dovitinib lactate
CAS 692737-80-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27FN6O4
Quantity 500 mg
Synonym CHIR-258 lactate TKI-258 lactate
Solubility Information DMSO : ≥ 30 mg/mL (62.17 mM)
SMILES O=C(N1)C(C(NC2=C3)=NC2=CC=C3N4CCN(C)CC4)=C(N)C5=C1C=CC=C5F.O=C(O)C(C)O
Molecular Weight (g/mol) 482.51
Formula Weight 482.51
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.