missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dovitinib lactate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Description
Dovitinib lactate (TKI258 lactate) is a multi-targeted tyrosine kinase inhibitor with IC50s of 1, 2, 8/9, 10/13/8, 27/210 nM for FLT3, c-Kit, FGFR1/3, VEGFR1/2/3 and PDGFRα/β, respectively.
Specifications
Specifications
| Chemical Name or Material | Dovitinib lactate |
| CAS | 692737-80-7 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H27FN6O4 |
| Quantity | 200 mg |
| Synonym | CHIR-258 lactate TKI-258 lactate |
| Solubility Information | DMSO : ≥ 30 mg/mL (62.17 mM) |
| SMILES | O=C(N1)C(C(NC2=C3)=NC2=CC=C3N4CCN(C)CC4)=C(N)C5=C1C=CC=C5F.O=C(O)C(C)O |
| Molecular Weight (g/mol) | 482.51 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction