missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxazosin, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Doxazosin
Specifications
Specifications
| Chemical Name or Material | Doxazosin |
| CAS | 74191-85-8 |
| Analyte or Component Names | Doxazosin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H25 N5 O5 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Doxazosin, 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(2,3-dihydro-1,4-benzodioxin-2-yl)carbonyl]piperazine, 4-Amino-2-[4-(1,4-benzodioxan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazoline, UK-33274 |
| InChI Formula | InChI=1S/C23H25N5O5/c1-30-18-11-14-15(12-19(18)31-2)25-23(26-21(14)24)28-9-7-27(8-10-28)22(29)20-13-32-16-5-3-4-6-17(16)33-20/h3-6,11-12,20H,7-10,13H2,1-2H3,(H2,24,25,26) |
| SMILES | COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4COc5ccccc5O4 |
| IUPAC Name | [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(2,3-dihydro-1,4-benzodioxin-3-yl)methanone |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction