Learn More
SREC-I/SCARF1 Antibody (373606), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2409A
Product Code. 18606973
  • ONLINE EXCLUSIVE PRICE
    € 664.00
    € 627.90 / 0.10mL
    Save € 36.10
    5% Off
Description
SREC-I/SCARF1 Monoclonal antibody specifically detects SREC-I/SCARF1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SREC-I/SCARF1
Monoclonal
APC
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
373606
Western Blot, Flow Cytometry, CyTOF-ready
Acetyl LDL receptor, KIAA0149endothelial cells scavenger receptor, MGC47738, scavenger receptor class F, member 1, Scavenger receptor expressed by endothelial cells 1, SREC1, SREC-I, SRECscavenger receptor expressed by endothelial cells
Mouse myeloma cell line NS0-derived recombinant human SREC-I/SCARF1, Ser20-Thr421, Accession # Q14162
0.1 mL
Lipid and Metabolism
8578
Store at 4°C in the dark.
IgG2b