Learn More
Invitrogen™ DP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527685
Description
Recommended positive controls: PC-3, mouse muscle. Predicted reactivity: Mouse (100%), Rat (97%), Zebrafish (84%), Xenopus laevis (86%), Chicken (95%), Rhesus Monkey (99%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The gene is a member of the transcription factor DP family. The encoded protein forms heterodimers with the E2F transcription factors resulting in transcriptional activation of cell cycle regulated genes. Alternative splicing results in multiple transcript variants.
Specifications
| DP2 | |
| Polyclonal | |
| Unconjugated | |
| TFDP2 | |
| 1110029I05Rik; A330080J22Rik; BTPS2; DP2; DP2.5; DP3; DP-3; E2F dimerization partner 2; GS; Tcfdp2; TFDP2; transcription factor Dp 2; transcription factor Dp-2; transcription factor Dp-2 (E2F dimerization partner 2); Unknown (protein for MGC:133813) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 211586, 7029 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.98 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14188, Q64163 | |
| TFDP2 | |
| Recombinant protein encompassing a sequence within the center region of human DP2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.