missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPCD Monoclonal Antibody (K1E042_4B5), Invitrogen™
Description
Sequence of this protein is as follows: RTAQKTALLQ DGRRKVHYLF PDGKEMAEEY DEKTSELLVR KWRVKSALGA MGQWQLEVGD PAPLGAGNLG PELIKESNAN PIFMRKDTKM SFQWRIRNLP YPKDVYSVSV DQKERCIIVR TTNKKYYKKF SIPDLDRHQL PLDDALLSFA HANCTLIISY QKPKEVVVAE SELQKELKKV KTAHSNDGDC
May play a role in the formation or function of ciliated cells.
Specifications
Specifications
| Antigen | DPCD |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | K1E042_4B5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | DPCD |
| Gene Accession No. | Q8BPA8, Q9BVM2 |
| Gene Alias | 5330431N19Rik; deleted in a mouse model of primary ciliary dyskinesia; deleted in primary ciliary dyskinesia; deleted in primary ciliary dyskinesia homolog (mouse); DPCD; Ndac; neighbor of Dac; protein DPCD; RGD1307648; RP11-529I10.4; RP11-529I104 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?