missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPCD Monoclonal Antibody (K1E042_4B5), Invitrogen™

Product Code. p-200088774
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30200326 200 μL 200µg
30209787 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30200326 Supplier Invitrogen™ Supplier No. MA556055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: RTAQKTALLQ DGRRKVHYLF PDGKEMAEEY DEKTSELLVR KWRVKSALGA MGQWQLEVGD PAPLGAGNLG PELIKESNAN PIFMRKDTKM SFQWRIRNLP YPKDVYSVSV DQKERCIIVR TTNKKYYKKF SIPDLDRHQL PLDDALLSFA HANCTLIISY QKPKEVVVAE SELQKELKKV KTAHSNDGDC

May play a role in the formation or function of ciliated cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPCD
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K1E042_4B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene DPCD
Gene Accession No. Q8BPA8, Q9BVM2
Gene Alias 5330431N19Rik; deleted in a mouse model of primary ciliary dyskinesia; deleted in primary ciliary dyskinesia; deleted in primary ciliary dyskinesia homolog (mouse); DPCD; Ndac; neighbor of Dac; protein DPCD; RGD1307648; RP11-529I10.4; RP11-529I104
Gene Symbols DPCD
Host Species Mouse
Immunogen Recombinant human DPCD (immunogen range is 11-200).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226162, 25911
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.